siRNA Details
Gene Symbol: RGD1559578 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134497.1 | NP_001127969.1 | 4, 5 | 704 |

Gene Symbol: RGD1559578 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134497.1 | NP_001127969.1 | 4, 5 | 704 |