This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RGD1308134 SIRNA Assays ›
Gene Symbol:
RGD1308134
Gene Name:
similar to RIKEN cDNA 1110020A23
Gene Aliases:
-
Chromosome Location:
Chr.10: 56843846 - 56848270 on Build Rnor_6.0
UniGene ID:
Rn.228770
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s142496

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RGD1308134
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127521.2 NP_001120993.1 4 348
XM_006246652.2 XP_006246714.1 4 516
XM_006246653.2 XP_006246715.1 3 414
XM_006246654.2 XP_006246716.1 4 516
XM_006246655.2 XP_006246717.1 3 414
XM_006246656.2 XP_006246718.1 3 414
XM_006246657.2 XP_006246719.1 3 414
GenBank mRNA BC158879.1 - 4 348 7936013
FQ228078.1 - 1 44

Back To Top

More Information


Gene Ontology Categories:

Process(es)

DNA binding
chromatin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0