siRNA Details
Gene Symbol: Ubtd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767629.1 | XP_008765851.1 | 4 | 526 |

Gene Symbol: Ubtd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767629.1 | XP_008765851.1 | 4 | 526 |