This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PDLIM7 SIRNA Assays ›
Gene Symbol:
Pdlim7
Gene Name:
PDZ and LIM domain 7
Gene Aliases:
Enigma
Chromosome Location:
Chr.17: 9653264 - 9668712 on Build Rnor_6.0
UniGene ID:
Rn.7274
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s141810

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pdlim7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173125.1 NP_775148.1 12, 13 1352
XM_006253610.2 XP_006253672.2 12, 13 1543
XM_008771487.1 XP_008769709.1 12, 13 1645
GenBank mRNA AF095585.1 - 12, 13 1352
BC078693.1 - 12, 13 1344 7121395

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

ossification
multicellular organism development
actin cytoskeleton organization
cell differentiation
positive regulation of osteoblast differentiation
protein binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0