This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PRL8A5 SIRNA Assays ›
Gene Symbol:
Prl8a5
Gene Name:
prolactin family 8, subfamily a, member 5
Gene Aliases:
-
Chromosome Location:
Chr.17: 39411716 - 39417929 on Build Rnor_6.0
UniGene ID:
Rn.3757
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s141771

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Prl8a5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173110.1 NP_775133.1 3, 4 289
XM_006253907.1 XP_006253969.1 3, 4 299
XM_006253908.1 XP_006253970.1 3, 4 249
GenBank mRNA BC097274.1 - 2, 3 236 7441946
BC158577.1 - 3, 4 283 7461869
M76537.1 - 3, 4 266
U09099.1 - 3, 4 289

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

female pregnancy
hormone activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0