This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CCR9 SIRNA Assays ›
Gene Symbol:
Ccr9
Gene Name:
chemokine (C-C motif) receptor 9
Gene Aliases:
-
Chromosome Location:
Chr.8: 132828091 - 132842130 on Build Rnor_6.0
UniGene ID:
Rn.91140
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s141722

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ccr9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172329.1 NP_758832.1 3 483
XM_006244179.2 XP_006244241.1 3 1289
GenBank mRNA AF458780.1 - 3 483
BC091115.1 - 3 497 7381553

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_031200, NM_001256369, NM_006641, AF145439, BC095516, and others.
This siRNA also targets the following transcripts in Mouse: NM_009913, NM_001166625, BC132466, AJ132336, AK019478, and others.

Gene Ontology Categories:

Function(s) Process(es)

chemotaxis
immune response
G-protein coupled receptor signaling pathway
chemokine-mediated signaling pathway
C-C chemokine receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0