This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PYCARD SIRNA Assays ›
Gene Symbol:
Pycard
Gene Name:
PYD and CARD domain containing
Gene Aliases:
Asc
Chromosome Location:
Chr.1: 199438029 - 199439062 on Build Rnor_6.0
UniGene ID:
Rn.7817
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s141694

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pycard
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172322.1 NP_758825.1 1 166
GenBank mRNA AB053165.1 - 1 166
FQ226280.1 - 1 324
FQ227544.1 - 1 345
FQ227755.1 - 1 324

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

myeloid dendritic cell activation
activation of innate immune response
positive regulation of defense response to virus by host
myeloid dendritic cell activation involved in immune response
positive regulation of antigen processing and presentation of peptide antigen via MHC class II
positive regulation of adaptive immune response
activation of cysteine-type endopeptidase activity involved in apoptotic process
inflammatory response
response to bacterium
regulation of autophagy
regulation of tumor necrosis factor-mediated signaling pathway
positive regulation of actin filament polymerization
regulation of protein stability
negative regulation of NF-kappaB transcription factor activity
interleukin-1 beta production
negative regulation of interferon-beta production
positive regulation of interferon-gamma production
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
tumor necrosis factor-mediated signaling pathway
positive regulation of activated T cell proliferation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
regulation of apoptotic process
positive regulation of apoptotic process
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of cysteine-type endopeptidase activity involved in apoptotic process
macropinocytosis
positive regulation of JNK cascade
positive regulation of interleukin-1 beta secretion
regulation of inflammatory response
positive regulation of phagocytosis
defense response to Gram-negative bacterium
positive regulation of T cell activation
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
defense response to virus
positive regulation of ERK1 and ERK2 cascade
cellular response to lipopolysaccharide
cellular response to interleukin-1
cellular response to tumor necrosis factor
negative regulation of protein serine/threonine kinase activity
intrinsic apoptotic signaling pathway by p53 class mediator
positive regulation of chemokine secretion
positive regulation of release of cytochrome c from mitochondria
positive regulation of T cell migration
positive regulation of interleukin-8 secretion
positive regulation of interleukin-6 secretion
positive regulation of cysteine-type endopeptidase activity
positive regulation of interleukin-10 secretion
positive regulation of extrinsic apoptotic signaling pathway
regulation of intrinsic apoptotic signaling pathway
protease binding
protein binding
Pyrin domain binding
protein homodimerization activity
ion channel binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0