This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PXMP4 SIRNA Assays ›
Gene Symbol:
Pxmp4
Gene Name:
peroxisomal membrane protein 4
Gene Aliases:
-
Chromosome Location:
Chr.3: 150091646 - 150108893 on Build Rnor_6.0
UniGene ID:
Rn.7752
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s141680

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pxmp4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172223.2 NP_757377.1 3 392
GenBank mRNA AY151834.1 - 3 370
BC081699.1 - 3 392 7193564

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_021534, AK003127, AK050647, AJ271476, AK156743, and others.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0