This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VPS4A SIRNA Assays ›
Gene Symbol:
Vps4a
Gene Name:
vacuolar protein sorting 4 homolog A (S. cerevisiae)
Gene Aliases:
-
Chromosome Location:
Chr.19: 39230024 - 39243298 on Build Rnor_6.0
UniGene ID:
Rn.12477
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s141062

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vps4a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145678.1 NP_663711.1 10 1100
XM_006255560.1 XP_006255622.1 10 1319
GenBank mRNA AB076398.1 - 10 1100
BC070931.1 - 10 1203 7096867
FQ213218.1 - 1280

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell separation after cytokinesis
nucleus organization
vacuole organization
mitotic metaphase plate congression
metabolic process
abscission
protein transport
vesicle-mediated transport
endosomal transport
cytoplasmic microtubule organization
negative regulation of cytokinesis
viral budding via host ESCRT complex
ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
cell division
ubiquitin-independent protein catabolic process via the multivesicular body sorting pathway
regulation of protein localization to plasma membrane
positive regulation of exosomal secretion
positive regulation of viral budding via host ESCRT complex
ATP binding
protein C-terminus binding
microtubule-severing ATPase activity
Rho GTPase binding
protein domain specific binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0