This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ARHGAP4 SIRNA Assays ›
Gene Symbol:
Arhgap4
Gene Name:
Rho GTPase activating protein 4
Gene Aliases:
-
Chromosome Location:
Chr.X: 156873100 - 156888762 on Build Rnor_6.0
UniGene ID:
Rn.163458
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s140887

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Arhgap4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144740.3 NP_653341.2 6 860
XM_006229545.2 XP_006229607.1 6 1620
XM_006229546.2 XP_006229608.1 5 1557
XM_006229547.2 XP_006229609.1 6 1620
XM_006229548.2 XP_006229610.1 6 1620
GenBank mRNA AF401635.1 - 6 870
AY742900.1 - 873

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal transduction
nervous system development
negative regulation of fibroblast migration
negative regulation of axon extension
regulation of Rho protein signal transduction
GTPase activator activity
Rac GTPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0