This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SIPA1L1 SIRNA Assays ›
Gene Symbol:
Sipa1l1
Gene Name:
signal-induced proliferation-associated 1 like 1
Gene Aliases:
SPAL, Spa-1, Spa1, Spar
Chromosome Location:
Chr.6: 106052212 - 106163249 on Build Rnor_6.0
UniGene ID:
Rn.10835
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s140829

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sipa1l1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139330.1 NP_647546.1 2 1353
GenBank mRNA AF026504.1 - 2 1353
AY043226.1 - 1 916

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_015556, AF090989, AB007900, AF090990, BC146777, and others.

Gene Ontology Categories:

Function(s) Process(es)

actin cytoskeleton reorganization
positive regulation of GTPase activity
ephrin receptor signaling pathway
regulation of synaptic plasticity
regulation of dendrite morphogenesis
regulation of axonogenesis
regulation of small GTPase mediated signal transduction
regulation of dendritic spine morphogenesis
GTPase activator activity
protein binding
protein kinase binding
ubiquitin protein ligase binding
protein complex binding
ephrin receptor binding
actin filament binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0