This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SCN3B SIRNA Assays ›
Gene Symbol:
Scn3b
Gene Name:
sodium channel, voltage-gated, type III, beta subunit
Gene Aliases:
Scnb3
Chromosome Location:
Chr.8: 44136414 - 44159011 on Build Rnor_6.0
UniGene ID:
Rn.3402
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s140661

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Scn3b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139097.3 NP_620797.1 3 497
XM_006243102.1 XP_006243164.1 3 579
XM_006243103.1 XP_006243165.1 3 579
XM_008766114.1 XP_008764336.1 3 579
XM_008766115.1 XP_008764337.1 2 146
GenBank mRNA BC070899.1 - 3 497 7100476

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

sodium ion transport
nervous system development
positive regulation of heart rate
positive regulation of sodium ion transport
sensory perception of pain
regulation of ion transmembrane transport
sodium ion transmembrane transport
membrane depolarization
cardiac muscle contraction
regulation of atrial cardiac muscle cell membrane depolarization
regulation of ventricular cardiac muscle cell membrane depolarization
cardiac conduction
protein localization to plasma membrane
cardiac muscle cell action potential involved in contraction
ventricular cardiac muscle cell action potential
membrane depolarization during action potential
membrane depolarization during cardiac muscle cell action potential
atrial cardiac muscle cell action potential
SA node cell to atrial cardiac muscle cell communication
regulation of heart rate by cardiac conduction
regulation of sodium ion transmembrane transporter activity
molecular_function
sodium channel regulator activity
ion channel binding
voltage-gated sodium channel activity involved in cardiac muscle cell action potential

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0