This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PIGL SIRNA Assays ›
Gene Symbol:
Pigl
Gene Name:
phosphatidylinositol glycan anchor biosynthesis, class L
Gene Aliases:
-
Chromosome Location:
Chr.10: 48774018 - 48831848 on Build Rnor_6.0
UniGene ID:
Rn.10774
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s140485

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pigl
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138901.2 NP_620256.1 5, 6 540
XM_008767767.1 XP_008765989.1 5, 6 536
GenBank mRNA BC074020.1 - 5, 6 540 7104008
D88364.1 - 5, 6 921

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

GPI anchor biosynthetic process
N-acetylglucosaminylphosphatidylinositol deacetylase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0