This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other POU3F3 SIRNA Assays ›
Gene Symbol:
Pou3f3
Gene Name:
POU class 3 homeobox 3
Gene Aliases:
Brn1, RHS1
Chromosome Location:
Chr.9: 49479014 - 49482246 on Build Rnor_6.0
UniGene ID:
Rn.11354
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s140311

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pou3f3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138837.1 NP_620192.1 1 1844
GenBank mRNA AJ001641.1 - 1 1844

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008900.

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
kidney development
transcription, DNA-templated
transcription from RNA polymerase II promoter
nervous system development
positive regulation of cell proliferation
positive regulation of gene expression
cerebral cortex radially oriented cell migration
forebrain ventricular zone progenitor cell division
negative regulation of apoptotic process
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
metanephric ascending thin limb development
metanephric macula densa development
metanephric thick ascending limb development
metanephric loop of Henle development
metanephric DCT cell differentiation
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
sequence-specific DNA binding
HMG box domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0