This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PDE9A SIRNA Assays ›
Gene Symbol:
Pde9a
Gene Name:
phosphodiesterase 9A
Gene Aliases:
-
Chromosome Location:
Chr.20: 10123626 - 10216325 on Build Rnor_6.0
UniGene ID:
Rn.20981
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s140283

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pde9a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138543.1 NP_612552.1 10 733
XM_008772793.1 XP_008771015.1 10 747
XM_008772794.1 XP_008771016.1 10 747
XM_008772795.1 XP_008771017.1 10 747
GenBank mRNA AF372654.1 - 10 733
AY145898.1 - 10 752
BC078733.1 - 10 674
BC161837.1 - 10 706 9029236

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

metabolic process
cGMP-mediated signaling
metal ion binding
3',5'-cyclic-GMP phosphodiesterase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0