This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DNM3 SIRNA Assays ›
Gene Symbol:
Dnm3
Gene Name:
dynamin 3
Gene Aliases:
dynamin-2
Chromosome Location:
Chr.13: 79906704 - 80379967 on Build Rnor_6.0
UniGene ID:
Rn.11191
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s140264

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dnm3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138538.1 NP_612547.1 14, 15 1706
GenBank mRNA AF201839.1 - 14, 15 1706

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

endocytosis
synapse assembly
metabolic process
sperm ejaculation
filopodium assembly
positive regulation of filopodium assembly
regulation of dendritic spine morphogenesis
negative regulation of dendritic spine morphogenesis
GTPase activity
protein binding
GTP binding
type 1 metabotropic glutamate receptor binding
type 5 metabotropic glutamate receptor binding
nitric-oxide synthase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm