This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EXOC2 SIRNA Assays ›
Gene Symbol:
Exoc2 LOC103689971
Gene Name:
exocyst complex component 2
exocyst complex component 2-like
Gene Aliases:
Rsec5, Sec5l1
-
Chromosome Location:
Chr.17: 34665810 - 34842266 on Build Rnor_6.0
Chromosome Location:
Chr.17: 35396224 - 35518693 on Build Rnor_6.0
UniGene ID:
Rn.2869
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s140131

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Exoc2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134414.2 NP_599241.2 16 1917
XM_006253891.1 XP_006253953.1 17 1989
XM_006253893.2 XP_006253955.1 16 1873
XM_006253895.2 XP_006253957.1 15 1801
GenBank mRNA AF032666.1 - 16 1855
CO396803.1 - 2 215
Gene Symbol: LOC103689971
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006253876.2 XP_006253938.1 1989
XM_006253878.2 XP_006253940.1 1872
XM_006253880.2 XP_006253942.1 1800
XM_006253881.2 XP_006253943.1 1863
XM_008771607.1 XP_008769829.1 1917

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_025588, BC049102, AK016532, BC048154, BC031207, and others.

Gene Ontology Categories:

Function(s) Process(es)

exocytosis
Golgi to plasma membrane transport
protein transport
protein binding
Ral GTPase binding
protein kinase binding
protein N-terminus binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0