This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MARC2 SIRNA Assays ›
Gene Symbol:
Marc2
Gene Name:
mitochondrial amidoxime reducing component 2
Gene Aliases:
Mg87, Mosc2, mARC1
Chromosome Location:
Chr.13: 102724266 - 102755460 on Build Rnor_6.0
UniGene ID:
Rn.11802
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s140124

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Marc2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134410.2 NP_599237.1 6 847
GenBank mRNA AF095741.1 - 6 848
BC061734.1 - 6 874 5598768
FQ211188.1 - 3 186

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

metabolic process
nitrate metabolic process
oxidation-reduction process
catalytic activity
nitrate reductase activity
oxidoreductase activity
molybdenum ion binding
pyridoxal phosphate binding
molybdopterin cofactor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header