siRNA Details
Gene Symbol: Marc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134410.2 | NP_599237.1 | 1, 2 | 292 | ||
GenBank mRNA | AF095741.1 | - | 1, 2 | 293 | ||
BC061734.1 | - | 1, 2 | 319 | 5598768 |

Gene Symbol: Marc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134410.2 | NP_599237.1 | 1, 2 | 292 | ||
GenBank mRNA | AF095741.1 | - | 1, 2 | 293 | ||
BC061734.1 | - | 1, 2 | 319 | 5598768 |