This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UGGT1 SIRNA Assays ›
Gene Symbol:
Uggt1
Gene Name:
UDP-glucose glycoprotein glucosyltransferase 1
Gene Aliases:
UGT1, Ugcgl1, Uggt, rUGT1
Chromosome Location:
Chr.9: 42692156 - 42805684 on Build Rnor_6.0
UniGene ID:
Rn.162227
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s139803

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Uggt1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133596.1 NP_598280.1 13, 14 1344
XM_006244734.2 XP_006244796.1 13, 14 1566
XM_006244735.2 XP_006244797.1 13, 14 1566
GenBank mRNA AF200359.1 - 13, 14 1344

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_198899, BC068283, AK087883, AK144381, AK050532, and others.

Gene Ontology Categories:

Function(s) Process(es)

protein folding
protein glycosylation
'de novo' posttranslational protein folding
UDP-glucosylation
UDP-glucose:glycoprotein glucosyltransferase activity
protein binding
unfolded protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0