This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OPA1 SIRNA Assays ›
Gene Symbol:
Opa1
Gene Name:
optic atrophy 1
Gene Aliases:
-
Chromosome Location:
Chr.11: 74717600 - 74793902 on Build Rnor_6.0
UniGene ID:
Rn.225901
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s139770

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Opa1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133585.3 NP_598269.3 17 1808
XM_006248497.1 XP_006248559.1 19 2072
XM_006248498.1 XP_006248560.1 18 2018
XM_006248499.1 XP_006248561.1 18 1961
GenBank mRNA AY510274.1 - 17 1863
BC111071.1 - 17 1808 7324017
U93197.1 - 1 14

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitochondrial genome maintenance
neural tube closure
apoptotic process
mitochondrion organization
inner mitochondrial membrane organization
aging
visual perception
mitochondrial fusion
metabolic process
positive regulation of mitochondrial fusion
positive regulation of neuron maturation
negative regulation of release of cytochrome c from mitochondria
cellular senescence
positive regulation of dendrite development
negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
negative regulation of intrinsic apoptotic signaling pathway
molecular_function
GTPase activity
GTP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0