This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PANK4 SIRNA Assays ›
Gene Symbol:
Pank4
Gene Name:
pantothenate kinase 4
Gene Aliases:
Fang1
Chromosome Location:
Chr.5: 172367087 - 172384077 on Build Rnor_6.0
UniGene ID:
Rn.17321
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s139610

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pank4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133531.2 NP_598215.1 16 1862
GenBank mRNA AF399873.1 - 16 1862
FQ223969.1 - 16 1869
FQ224186.1 - 16 1871

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_018216, BC043496, AL442072, AK295301, AK223546, and others.
This siRNA also targets the following transcripts in Mouse: NM_172990, BC050089, AK050894.

Gene Ontology Categories:

Function(s) Process(es)

coenzyme A biosynthetic process
phosphorylation
pantothenate kinase activity
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header