This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MMP13 SIRNA Assays ›
Gene Symbol:
Mmp13
Gene Name:
matrix metallopeptidase 13
Gene Aliases:
-
Chromosome Location:
Chr.8: 5522739 - 5533018 on Build Rnor_6.0
UniGene ID:
Rn.10997
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s139606

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mmp13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133530.1 NP_598214.1 1, 2 139
GenBank mRNA M60616.1 - 1, 2 112

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008607, AK160819, BC125320, AK152507, AK159921, and others.

Gene Ontology Categories:

Function(s) Process(es)

ossification
luteolysis
response to hypoxia
endochondral ossification
growth plate cartilage development
proteolysis
response to mechanical stimulus
extracellular matrix disassembly
bone mineralization
collagen catabolic process
receptor internalization
embryonic hindlimb morphogenesis
response to drug
peptide catabolic process
response to estrogen
cellular protein metabolic process
cartilage development
bone morphogenesis
fibronectin binding
metalloendopeptidase activity
calcium ion binding
collagen binding
peptidase activity
zinc ion binding
calcium-dependent protein binding
low-density lipoprotein particle receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header