This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FOLR1 SIRNA Assays ›
Gene Symbol:
Folr1
Gene Name:
folate receptor 1 (adult)
Gene Aliases:
Fbp1
Chromosome Location:
Chr.1: 166934457 - 166943592 on Build Rnor_6.0
UniGene ID:
Rn.211829
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s139596

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Folr1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133527.1 NP_598211.1 4 406
XM_006229842.2 XP_006229904.1 4 527
GenBank mRNA AF219904.1 - 4 406
BC157811.1 - 3 389

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001252552, NM_001252553, NM_008034, NM_001252554, M64782, and others.

Gene Ontology Categories:

Function(s) Process(es)

folic acid transport
folic acid metabolic process
folic acid binding
folic acid transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0