This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RAP2B SIRNA Assays ›
Gene Symbol:
Rap2b
Gene Name:
RAP2B, member of RAS oncogene family
Gene Aliases:
-
Chromosome Location:
Chr.2: 151685251 - 151685802 on Build Rnor_6.0
UniGene ID:
Rn.217110
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s139500

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rap2b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133410.1 NP_596901.1 1 480
GenBank mRNA AF386786.1 - 1 480

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_028712, AK150187, AK014462, AK042941, BC046528, and others.

Gene Ontology Categories:

Function(s) Process(es)

microvillus assembly
platelet activation
negative regulation of cell migration
positive regulation of protein autophosphorylation
Rap protein signal transduction
regulation of protein tyrosine kinase activity
platelet aggregation
establishment of endothelial intestinal barrier
GTP binding
GDP binding
protein domain specific binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0