This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZDHHC7 SIRNA Assays ›
Gene Symbol:
Zdhhc7
Gene Name:
zinc finger, DHHC-type containing 7
Gene Aliases:
Serz-1, Serz1
Chromosome Location:
Chr.19: 52733161 - 52750307 on Build Rnor_6.0
UniGene ID:
Rn.7790
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s139452

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zdhhc7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133394.1 NP_596885.1 7 953
GenBank mRNA AY040615.1 - 7 953
AY886525.1 - 7 948
BC061769.1 - 6 729 5621307

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_133967, AK089681, BC029666, AY166673, AK213858, and others.

Gene Ontology Categories:

Function(s) Process(es)

peptidyl-L-cysteine S-palmitoylation
protein palmitoylation
zinc ion binding
palmitoyltransferase activity
protein-cysteine S-palmitoyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0