This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ICMT SIRNA Assays ›
Gene Symbol:
Icmt
Gene Name:
isoprenylcysteine carboxyl methyltransferase
Gene Aliases:
fcmt
Chromosome Location:
Chr.5: 169475270 - 169482031 on Build Rnor_6.0
UniGene ID:
Rn.48709
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s139373

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Icmt
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133310.2 NP_579844.2 3 374
GenBank mRNA AF075595.1 - 3 162
BC083923.1 - 4 579
CB616023.1 - 3 374

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
liver development
protein methylation
C-terminal protein methylation
protein localization
positive regulation of cell proliferation
multicellular organism growth
S-adenosylhomocysteine metabolic process
S-adenosylmethioninamine metabolic process
regulation of Ras protein signal transduction
regulation of nucleic acid-templated transcription
regulation of RNA biosynthetic process
protein C-terminal S-isoprenylcysteine carboxyl O-methyltransferase activity
cAMP response element binding protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0