This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BAIAP2 SIRNA Assays ›
Gene Symbol:
Baiap2
Gene Name:
BAI1-associated protein 2
Gene Aliases:
Irsp53
Chromosome Location:
Chr.10: 109107326 - 109187458 on Build Rnor_6.0
UniGene ID:
Rn.95155
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s138806

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Baiap2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_057196.1 NP_476544.1 12 1439
XM_006247869.2 XP_006247931.1 12 1502
XM_006247870.2 XP_006247932.1 12 1502
XM_006247871.2 XP_006247933.1 12 1382
XM_006247872.2 XP_006247934.1 12 1382
XM_008768464.1 XP_008766686.1 12 1502
GenBank mRNA AY037934.1 - 12 1439
BC074009.1 - 12 1473 7099884

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

plasma membrane organization
regulation of cell shape
response to bacterium
dendrite development
regulation of actin cytoskeleton organization
regulation of synaptic plasticity
actin filament bundle assembly
actin crosslink formation
positive regulation of dendritic spine morphogenesis
protein binding
cytoskeletal adaptor activity
PDZ domain binding
identical protein binding
proline-rich region binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0