This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other HEPH SIRNA Assays ›
Gene Symbol:
Heph
Gene Name:
hephaestin
Gene Aliases:
-
Chromosome Location:
Chr.X: 65377056 - 65658479 on Build Rnor_6.0
UniGene ID:
Rn.43849
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s138622

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Heph
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133304.1 NP_579838.1 8 1343
XM_006257046.2 XP_006257108.1 9 1643
XM_006257048.2 XP_006257110.1 9 1878
XM_006257049.2 XP_006257111.1 8 1678
XM_008773248.1 XP_008771470.1 10 2276
XM_008773249.1 XP_008771471.1 10 1972
XM_008773250.1 XP_008771472.1 9 1646
GenBank mRNA AF246120.1 - 8 1343

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010417, NM_181273, NM_001159627, NM_001159628, BC048237, and others.

Gene Ontology Categories:

Function(s) Process(es)

copper ion transport
iron ion transport
cellular iron ion homeostasis
erythrocyte differentiation
oxidation-reduction process
ferroxidase activity
copper ion binding
ferrous iron binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0