This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UBR5 SIRNA Assays ›
Gene Symbol:
Ubr5
Gene Name:
ubiquitin protein ligase E3 component n-recognin 5
Gene Aliases:
Dd5, Rat100
Chromosome Location:
Chr.7: 76789110 - 76835518 on Build Rnor_6.0
UniGene ID:
Rn.54812
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s138525

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ubr5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008765482.1 XP_008763704.1 9 1238

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
DNA repair
cellular response to DNA damage stimulus
protein ubiquitination
positive regulation of protein import into nucleus, translocation
positive regulation of catenin import into nucleus
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
progesterone receptor signaling pathway
positive regulation of canonical Wnt signaling pathway
negative regulation of histone H2A K63-linked ubiquitination
negative regulation of double-strand break repair
RNA binding
ubiquitin-protein transferase activity
zinc ion binding
ligase activity
protein domain specific binding
ubiquitin-ubiquitin ligase activity
ubiquitin binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0