This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other POU2F2 SIRNA Assays ›
Gene Symbol:
Pou2f2
Gene Name:
POU class 2 homeobox 2
Gene Aliases:
2-Oct, Oct2
Chromosome Location:
Chr.1: 81966760 - 82003740 on Build Rnor_6.0
UniGene ID:
Rn.155998
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s138521

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pou2f2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006228368.2 XP_006228430.2 3 161
XM_006228371.2 XP_006228433.2 3 161
XM_006228372.2 XP_006228434.2 3 161
XM_008758885.1 XP_008757107.1 3 161
GenBank mRNA L25863.1 - 3 112

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001163556, NM_001163554, NM_001163555, NM_011138, X57940, and others.

Gene Ontology Categories:

Function(s) Process(es)

mature B cell differentiation
immunoglobulin secretion involved in immune response
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
immunoglobulin secretion
cell maturation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein domain specific binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0