This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EIF4E SIRNA Assays ›
Gene Symbol:
Eif4e
Gene Name:
eukaryotic translation initiation factor 4E
Gene Aliases:
-
Chromosome Location:
Chr.2: 243819614 - 243853987 on Build Rnor_6.0
UniGene ID:
Rn.11275
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s138487

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Eif4e
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053974.2 NP_446426.1 7 578
XM_008761488.1 XP_008759710.1 8 691
GenBank mRNA BC087001.1 - 7 578 7122606
FQ218941.1 - 7 570
FQ225918.1 - 2 70
X83399.1 - 7 613

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007917, AK146757, BC010759, M61731, AK167097, and others.

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
behavioral fear response
translation
translational initiation
regulation of translation
lung development
positive regulation of mitotic cell cycle
translation initiation factor activity
protein binding
eukaryotic initiation factor 4G binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0