This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RRAGA SIRNA Assays ›
Gene Symbol:
Rraga
Gene Name:
Ras-related GTP binding A
Gene Aliases:
Raga
Chromosome Location:
Chr.5: 104941067 - 104942662 on Build Rnor_6.0
UniGene ID:
Rn.2816
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s138485

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rraga
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053973.2 NP_446425.1 1 469
GenBank mRNA BC061850.1 - 1 470 5621998
CK599679.1 - 1 469
X85183.1 - 1 454

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_178376, BC048245, BC037615, AK144591, AK004955.

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
biological_process
metabolic process
cell death
cellular response to starvation
regulation of autophagy
modulation by virus of host morphology or physiology
positive regulation of TOR signaling
cellular protein localization
positive regulation of cytolysis
cellular response to amino acid stimulus
GTPase activity
GTP binding
GDP binding
protein homodimerization activity
protein heterodimerization activity
phosphoprotein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0