This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PTF1A SIRNA Assays ›
Gene Symbol:
Ptf1a
Gene Name:
pancreas specific transcription factor, 1a
Gene Aliases:
-
Chromosome Location:
Chr.17: 86199623 - 86201477 on Build Rnor_6.0
UniGene ID:
Rn.10536
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s138460

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ptf1a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053964.1 NP_446416.1 1 841
GenBank mRNA X98170.1 - 1 833

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
tissue development
retina layer formation
cerebellum development
cell differentiation
hindbrain development
pancreas development
exocrine pancreas development
amacrine cell differentiation
cell fate commitment
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
retinoic acid receptor signaling pathway
neuron fate commitment
generation of neurons
retina morphogenesis in camera-type eye
regulation of neural retina development
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
protein dimerization activity
E-box binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0