This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other APBB3 SIRNA Assays ›
Gene Symbol:
Apbb3
Gene Name:
amyloid beta (A4) precursor protein-binding, family B, member 3
Gene Aliases:
Fe65l2
Chromosome Location:
Chr.18: 29497865 - 29504995 on Build Rnor_6.0
UniGene ID:
Rn.30067
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s138441

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Apbb3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053957.1 NP_446409.1 3, 4 569
XM_006254495.1 XP_006254557.1 3, 4 569
GenBank mRNA BC097935.1 - 3, 4 549
Y13413.1 - 3, 4 569

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
beta-amyloid binding
protein binding
transcription factor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0