This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PCDHA13 SIRNA Assays ›
Gene Symbol:
Pcdha13 LOC317356
Gene Name:
protocadherin alpha 13
similar to protocadherin alpha 13
Gene Aliases:
Cnr5
-
Chromosome Location:
Chr.18: 30036887 - 30215897 on Build Rnor_6.0
Chromosome Location:
Chr.X: 13709703 - 13713872 on Build Rnor_6.0
UniGene ID:
Rn.22308
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s138365

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pcdha13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053934.2 NP_446386.2 1 821
GenBank mRNA AB045586.1 - 1 99
AB113396.1 - 1 821
AY573975.1 - 1 821
Gene Symbol: LOC317356
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006256688.1 XP_006256750.1 2 245

Back To Top

More Information


Gene Ontology Categories:

Function(s)

odontogenesis

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0