This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MPRIP SIRNA Assays ›
Gene Symbol:
Mprip
Gene Name:
myosin phosphatase Rho interacting protein
Gene Aliases:
M-rip, Mrip, Rhoip3, p116Rip
Chromosome Location:
Chr.10: 46018390 - 46133579 on Build Rnor_6.0
UniGene ID:
Rn.2641
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s138011

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mprip
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034022.1 NP_001029194.1 4, 5 489
NM_053814.2 NP_446266.2 4, 5 489
XM_006246422.2 XP_006246484.1 4, 5 757
XM_006246423.2 XP_006246485.1 4, 5 757
XM_006246424.2 XP_006246486.1 4, 5 757
XM_006246425.2 XP_006246487.1 4, 5 756
XM_006246427.2 XP_006246489.1 4, 5 757
XM_006246431.2 XP_006246493.1 4, 5 744
XM_006246432.2 XP_006246494.1 4, 5 742
XM_008767759.1 XP_008765981.1 4, 5 757
XM_008767760.1 XP_008765982.1 4, 5 757
XM_008767761.1 XP_008765983.1 4, 5 758
XM_008767762.1 XP_008765984.1 4, 5 744
GenBank mRNA AF311311.1 - 4, 5 419
BC098911.1 - 4, 5 475 7446349
FQ222001.1 - 4, 5 527

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_012027, NM_201245, BC049803, AK158296, AK162530, and others.

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
maintenance of protein location in cell
negative regulation of myosin-light-chain-phosphatase activity
molecular_function
actin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0