This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GALE SIRNA Assays ›
Gene Symbol:
Gale
Gene Name:
UDP-galactose-4-epimerase
Gene Aliases:
-
Chromosome Location:
Chr.5: 154310453 - 154314959 on Build Rnor_6.0
UniGene ID:
Rn.29908
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s137882

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gale
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080783.2 NP_542961.2 7, 8 744
XM_006239134.2 XP_006239196.1 7, 8 1709
XM_008764197.1 XP_008762419.1 7, 8 975
GenBank mRNA BC097293.1 - 7, 8 744 7443707
X53949.1 - 6, 7 722

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

galactose metabolic process
galactose catabolic process
UDP-N-acetylglucosamine 4-epimerase activity
UDP-glucose 4-epimerase activity
protein homodimerization activity
coenzyme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0