This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DUSP1 SIRNA Assays ›
Gene Symbol:
Dusp1
Gene Name:
dual specificity phosphatase 1
Gene Aliases:
3CH134, CL100, MKP-1, Mkp1, Ptpn16
Chromosome Location:
Chr.10: 16970642 - 16973425 on Build Rnor_6.0
UniGene ID:
Rn.98260
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s137874

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dusp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053769.3 NP_446221.2 3 862
GenBank mRNA S81478.1 - 3 836
U02553.1 - 3 848
X84004.1 - 3 885

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

inactivation of MAPK activity
endoderm formation
protein dephosphorylation
response to light stimulus
response to organic substance
response to estradiol
response to retinoic acid
cellular response to hormone stimulus
response to testosterone
peptidyl-tyrosine dephosphorylation
intracellular signal transduction
peptidyl-threonine dephosphorylation
response to hydrogen peroxide
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of MAP kinase activity
negative regulation of MAPK cascade
response to glucocorticoid
negative regulation of meiotic cell cycle
response to cAMP
response to calcium ion
negative regulation of ERK1 and ERK2 cascade
mitotic cell cycle arrest
regulation of mitotic cell cycle spindle assembly checkpoint
negative regulation of DNA biosynthetic process
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
protein tyrosine/threonine phosphatase activity
MAP kinase tyrosine/serine/threonine phosphatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0