This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ITGA6 SIRNA Assays ›
Gene Symbol:
Itga6
Gene Name:
integrin, alpha 6
Gene Aliases:
-
Chromosome Location:
Chr.3: 58442904 - 58515124 on Build Rnor_6.0
UniGene ID:
Rn.22382
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s137707

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Itga6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053725.1 NP_446177.1 10, 11 1682
XM_006234351.1 XP_006234413.1 10, 11 1879
GenBank mRNA AJ312934.1 - 8, 9 1192

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008397, NM_001277970, AK045391, BC058095, AK159653, and others.

Gene Ontology Categories:

Function(s) Process(es)

cell-matrix adhesion
integrin-mediated signaling pathway
ectodermal cell differentiation
positive regulation of cell-substrate adhesion
single organismal cell-cell adhesion
positive regulation of cell-cell adhesion
cell-substrate adhesion
cellular response to extracellular stimulus
cell adhesion mediated by integrin
nail development
positive regulation of phosphorylation
odontogenesis of dentin-containing tooth
positive regulation of apoptotic process
skin development
positive regulation of transcription from RNA polymerase II promoter
filopodium assembly
digestive tract development
brown fat cell differentiation
leukocyte migration
cellular response to organic cyclic compound
renal system development
amelogenesis
negative regulation of extrinsic apoptotic signaling pathway
integrin binding
protein complex binding
laminin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0