This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EBNA1BP2 SIRNA Assays ›
Gene Symbol:
Ebna1bp2 LOC100912375
Gene Name:
EBNA1 binding protein 2
probable rRNA-processing protein EBP2-like
Gene Aliases:
Ebp2
-
Chromosome Location:
Chr.5: 137670165 - 137674730 on Build Rnor_6.0
Chromosome Location:
Chr.5: 137458672 - 137463255 on Build Rnor_6.0
UniGene ID:
Rn.9598
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID s137414

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ebna1bp2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008721.1 NP_001008721.1 806
GenBank mRNA BC087738.1 - 806 7310934
DQ480746.1 - 1796
FQ213511.1 - 829
FQ219880.1 - 751
FQ231568.1 - 828
Gene Symbol: LOC100912375
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003750007.3 XP_003750055.1 8 826

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026932, AK007491, BC062876, AK168269, AK167070, and others.

Gene Ontology Categories:

Process(es)

poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0