This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C5AR1 SIRNA Assays ›
Gene Symbol:
C5ar1
Gene Name:
complement component 5a receptor 1
Gene Aliases:
C5r1
Chromosome Location:
Chr.1: 78186777 - 78195132 on Build Rnor_6.0
UniGene ID:
Rn.10680
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s137366

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C5ar1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053619.1 NP_446071.1 2 335
GenBank mRNA AB003042.1 - 2 335
AF090996.1 - 1 50
BC078770.1 - 2 318 7121456
Y09613.1 - 2 319

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

complement receptor mediated signaling pathway
apoptotic process
inflammatory response
G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
cell proliferation in hindbrain
neutrophil chemotaxis
organ regeneration
response to peptidoglycan
response to lipopolysaccharide
complement component C5a signaling pathway
mRNA transcription from RNA polymerase II promoter
negative regulation of neuron apoptotic process
positive regulation of epithelial cell proliferation
defense response to Gram-positive bacterium
positive regulation of ERK1 and ERK2 cascade
complement component C5a binding
complement component C5a receptor activity
C5a anaphylatoxin receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0