This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNCB SIRNA Assays ›
Gene Symbol:
Sncb
Gene Name:
synuclein, beta
Gene Aliases:
-
Chromosome Location:
Chr.17: 10384472 - 10392776 on Build Rnor_6.0
UniGene ID:
Rn.20352
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s137285

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sncb
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080777.2 NP_542955.2 4 388
XM_006253594.2 XP_006253656.1 5 589
GenBank mRNA BC087579.1 - 4 388 7314241
D17764.1 - 4 244

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chemical synaptic transmission
dopamine metabolic process
negative regulation of neuron apoptotic process
synapse organization
calcium ion binding
alpha-tubulin binding
beta-tubulin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header