This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TINAGL1 SIRNA Assays ›
Gene Symbol:
Tinagl1
Gene Name:
tubulointerstitial nephritis antigen-like 1
Gene Aliases:
Lcn7, gis5
Chromosome Location:
Chr.5: 148382702 - 148392719 on Build Rnor_6.0
UniGene ID:
Rn.1256
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s137193

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tinagl1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053582.2 NP_446034.1 4 563
XM_006238977.2 XP_006239039.1 5 655
XM_006238978.2 XP_006239040.1 5 652
GenBank mRNA AB050717.1 - 4 564
BC097299.1 - 5 622 7445067

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
receptor-mediated endocytosis
immune response
scavenger receptor activity
cysteine-type peptidase activity
polysaccharide binding
laminin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0