This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NAP1L1 SIRNA Assays ›
Gene Symbol:
Nap1l1
Gene Name:
nucleosome assembly protein 1-like 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 54213305 - 54237880 on Build Rnor_6.0
UniGene ID:
Rn.162938
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s137127

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nap1l1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053561.2 NP_446013.2 7, 8 731
XM_006241342.1 XP_006241404.1 7, 8 745
XM_006241343.2 XP_006241405.1 7, 8 745
XM_006241344.2 XP_006241406.1 5, 6 482
GenBank mRNA AF062594.1 - 7, 8 713
CF110129.1 - 2, 3 166

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

nucleosome assembly
Schwann cell proliferation
cellular response to peptide hormone stimulus
kinase binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0