This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC2A8 SIRNA Assays ›
Gene Symbol:
Slc2a8
Gene Name:
solute carrier family 2, (facilitated glucose transporter) member 8
Gene Aliases:
Glut8
Chromosome Location:
Chr.3: 12020227 - 12029918 on Build Rnor_6.0
UniGene ID:
Rn.12923
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s136880

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc2a8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053494.2 NP_445946.2 7 979
XM_008761779.1 XP_008760001.1 7 1020
GenBank mRNA AJ245935.1 - 7 979

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
male meiosis I
insulin receptor signaling pathway
glucose transport
hexose transmembrane transport
transmembrane transport
glucose transmembrane transporter activity
glucose binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0