This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PLG SIRNA Assays ›
Gene Symbol:
Plg
Gene Name:
plasminogen
Gene Aliases:
Ab1-346
Chromosome Location:
Chr.1: 48521830 - 48563759 on Build Rnor_6.0
UniGene ID:
Rn.20178
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s136871

Purification
Size
Availability Made To Order
Catalog # 4390771
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Plg
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053491.2 NP_445943.1 16 1963
GenBank mRNA AJ242649.1 - 16 1966
BC091135.1 - 16 1963 7369357
FQ210878.1 - 16 2106
FQ218248.1 - 3 206

Back To Top

More Information


Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

blood coagulation
negative regulation of cell-substrate adhesion
extracellular matrix disassembly
tissue regeneration
fibrinolysis
myoblast differentiation
muscle cell cellular homeostasis
tissue remodeling
proteolysis involved in cellular protein catabolic process
negative regulation of fibrinolysis
positive regulation of fibrinolysis
trophoblast giant cell differentiation
labyrinthine layer blood vessel development
mononuclear cell migration
endopeptidase activity
serine-type endopeptidase activity
receptor binding
protein domain specific binding
apolipoprotein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0