This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SEMA6B SIRNA Assays ›
Gene Symbol:
Sema6b
Gene Name:
sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6B
Gene Aliases:
-
Chromosome Location:
Chr.9: 10934432 - 10951252 on Build Rnor_6.0
UniGene ID:
Rn.87343
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s136812

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sema6b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053471.2 NP_445923.1 3, 4 518
XM_006244348.2 XP_006244410.1 4, 5 987
XM_006244349.2 XP_006244411.1 3, 4 340
GenBank mRNA AB000776.1 - 3, 4 518

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

nervous system development
cell differentiation
receptor activity
semaphorin receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0