This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NCOA3 SIRNA Assays ›
Gene Symbol:
Ncoa3
Gene Name:
nuclear receptor coactivator 3
Gene Aliases:
Aib1, Tram-1
Chromosome Location:
Chr.3: 162692176 - 162788582 on Build Rnor_6.0
UniGene ID:
Rn.20691
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s136752

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ncoa3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006235634.1 XP_006235696.1 5 496

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
signal transduction
male gonad development
positive regulation of gene expression
histone acetylation
positive regulation of cell growth
cellular response to hormone stimulus
positive regulation of intracellular steroid hormone receptor signaling pathway
positive regulation of intracellular estrogen receptor signaling pathway
multicellular organism growth
negative regulation of apoptotic process
positive regulation of keratinocyte differentiation
positive regulation of transcription from RNA polymerase II promoter
developmental growth
positive regulation of sequence-specific DNA binding transcription factor activity
vagina development
labyrinthine layer morphogenesis
mammary gland branching involved in thelarche
regulation of RNA biosynthetic process
RNA polymerase II regulatory region DNA binding
chromatin binding
transcription coactivator activity
histone acetyltransferase activity
signal transducer activity
ligand-dependent nuclear receptor binding
estrogen receptor binding
ligand-dependent nuclear receptor transcription coactivator activity
protein complex binding
nuclear hormone receptor binding
peroxisome proliferator activated receptor binding
protein dimerization activity
protein N-terminus binding
androgen receptor binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0