This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GAB2 SIRNA Assays ›
Gene Symbol:
Gab2
Gene Name:
GRB2-associated binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 162083038 - 162282707 on Build Rnor_6.0
UniGene ID:
Rn.211990
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID s136641

Purification
Size
Availability Made To Order
Catalog # 4390771
Price 368.00
Your Price
Online offer: 309.12
309.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gab2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053417.1 NP_445869.1 2 362
XM_008759806.1 XP_008758028.1 1 1994
GenBank mRNA AF230367.2 - 2 362

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transmembrane receptor protein tyrosine kinase signaling pathway
integrin-mediated signaling pathway
positive regulation of cell proliferation
cell migration
osteoclast differentiation
intracellular signal transduction
positive regulation of mast cell degranulation
phosphatidylinositol-mediated signaling
transmembrane receptor protein tyrosine kinase adaptor activity
phosphatidylinositol-3,4,5-trisphosphate binding
protein domain specific binding
phosphatidylinositol-3,4-bisphosphate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0